EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
TRAF5-mediated Lys-63-linked Polyubiquitination Plays an Essential Role in Positive Regulation of ROR?t in Promoting IL-17A Expression.
PMID 26453305