EMAIL THIS PAGE TO A FRIEND
Environment International
Persistent and emerging pollutants in the blood of German adults: Occurrence of dechloranes, polychlorinated naphthalenes, and siloxanes.
PMID 26453819
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
370908
Tris(trimethylsiloxy)silane, ≥98%
C9H28O3Si4