EMAIL THIS PAGE TO A FRIEND
Carbohydrate Polymers
Anti-crease finishing of cotton fabrics based on crosslinking of cellulose with acryloyl malic acid.
PMID 26453855
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
549797
Acryloyl chloride, 97.0%, contains <210 ppm MEHQ as stabilizer
C3H3ClO
A24109
Acryloyl chloride, ≥97%, contains ~400 ppm phenothiazine as stabilizer
C3H3ClO
240176
DL-Malic acid, ReagentPlus®, ≥99%
C4H6O5
M0875
DL-Malic acid, ≥98% (capillary GC)
C4H6O5
27606
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
C4H6O5
W265501
DL-Malic acid, ≥99%
C4H6O5
M0375
Maleic acid, ReagentPlus®, ≥99.0% (HPLC)
C4H4O4
M8304
Malic acid, meets USP/NF testing specifications
C4H6O5
746444
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
BrK
793604
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
BrK
P0838
Potassium bromide, BioXtra, ≥99.0%
BrK
449962
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
BrK
451010
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
BrK
449970
Potassium bromide, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
BrK
229792
Potassium bromide, 99.99% trace metals basis
BrK
243418
Potassium bromide, ACS reagent, ≥99.0%
BrK
P9881
Potassium bromide, ReagentPlus®, ≥99.0%
BrK
221864
Potassium bromide, FT-IR grade, ≥99% trace metals basis
BrK
60089
Potassium bromide, BioUltra, ≥99.5% (AT)
BrK
60093
Potassium bromide, puriss. p.a., ACS reagent, ≥99.5% (AT)
BrK
02110
Potassium bromide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5%
BrK
04434
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%
H2NaO2P