EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Synthesis and characterization of a novel mucoadhesive derivative of xyloglucan.
PMID 26453888
Abstract
A novel polymer in the form of a thiolated derivative of natural tamarind seed polysaccharide or xyloglucan was synthesized and its chacteristics as a mucoadhesive polymer were studied as a part of the study undertaken herein. The synthetic route followed involves a two-step reaction mechanism of firstly oxidizing xyloglucan and then further conjugating it with l-cysteine to form thiolated xyloglucan or thiomer via imine linkage. The thiomer thus formed was characterized using various analytical techniques as differential scanning calorimetry (DSC), X-ray diffraction analysis (XRD), and nuclear magnetic resonance (NMR). Ellman's method was used to determine the numbers of thiol groups/g of thiolated xyloglucan. Zeta potential measurements were carried out for thiolated xyloglucan. Viscosities of the formulated xyloglucan and thiolated xyloglucan gels were comparatively evaluated along with the evaluation of mucoadhesive properties of the gels using ex vivo bioadhesion study employing freshly excised sheep intestinal mucosa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
168149
L-Cysteine, 97%
C3H7NO2S
C7352
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
C3H7NO2S
W326305
L-Cysteine, ≥97%, FG
C3H7NO2S
30089
L-Cysteine, BioUltra, ≥98.5% (RT)
C3H7NO2S
C5360
L-Cysteine, PharmaGrade, Ajinomoto, from non-animal source, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S
778672
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
C3H7NO2S
S1878
Sodium (meta)periodate, ≥99.0%
INaO4
71859
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
INaO4
30323
Sodium (meta)periodate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8%
INaO4
452882
Sodium borohydride, powder, ≥98.0%
H4BNa
213462
Sodium borohydride, ReagentPlus®, 99%
H4BNa
71320
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
H4BNa
480886
Sodium borohydride, granular, 99.99% trace metals basis
H4BNa
452874
Sodium borohydride, granular, 10-40 mesh, 98%
H4BNa
452890
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
H4BNa
247677
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
H4BNa
686018
Sodium borohydride, hydrogen-storage grade, 98%
H4BNa
452904
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
H4BNa
215511
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
H4BNa
200972
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
H4BNa
311448
Sodium periodate, ACS reagent, ≥99.8%
INaO4
363642
Sodium periodate, 10 wt. %, matrix silica gel support
INaO4
632287
VenPure® SF, powder
H4BNa