EMAIL THIS PAGE TO A FRIEND
Chemosphere
The relationship between prenatal exposure to BP-3 and Hirschsprung's disease.
PMID 26454118
Abstract
Hirschsprung's disease (HSCR) is neonatal intestinal abnormality which derived from the faliure of enteric neural crest cells migration to hindgut during embryogenesis from 5 to 12 weeks. Currenly, the knowledge of environmental factors contributing to HSCR is still scarce. Benzophenone-3 (BP-3) is one of the most widely used UV filters, and has weak estrogen and strong anti-androgenic effects. In order to examine the effect of maternal BP-3 exposure on development of offspring and explore the potential mechanism, we conducted case and control study and in vitro study. In this work, BP-3 concertrations in maternal urine was detected by ultra-high performance liquid chromatography. Besides, we investigated the cytotoxicity and receptor tyrosine kinase (RET) expression in cells exposed to BP-3. The results showed that maternal BP-3 exposure was associated with offspring's HSCR in the population as well as inhibited migration of 293T and SH-SY5Y cells. What's more, we discovered dose-response relationship between RET expression and BP-3 exposure dose, and miR-218 and some other genes involved in SLIT2/ROBO1-miR-218-RET/PLAG1 pathway were also related to BP-3 exposure. Therefore, we deduced that BP-3 influenced cell migration via SLIT2/ROBO1-miR-218-RET/PLAG1 pathway. Our study firstly revealed the relationship between maternal BP-3 exposure and HSCR as well as its potential mechanism.