EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
PERK pathway is involved in oxygen-glucose-serum deprivation-induced NF-kB activation via ROS generation in spinal cord astrocytes.
PMID 26454173