EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
A fluorescence high throughput screening method for the detection of reactive electrophiles as potential skin sensitizers.
PMID 26455772
Abstract
Skin sensitization is an important toxicological end-point in the risk assessment of chemical allergens. Because of the complexity of the biological mechanisms associated with skin sensitization, integrated approaches combining different chemical, biological and in silico methods are recommended to replace conventional animal tests. Chemical methods are intended to characterize the potential of a sensitizer to induce earlier molecular initiating events. The presence of an electrophilic mechanistic domain is considered one of the essential chemical features to covalently bind to the biological target and induce further haptenation processes. Current in chemico assays rely on the quantification of unreacted model nucleophiles after incubation with the candidate sensitizer. In the current study, a new fluorescence-based method, 'HTS-DCYA assay', is proposed. The assay aims at the identification of reactive electrophiles based on their chemical reactivity toward a model fluorescent thiol. The reaction workflow enabled the development of a High Throughput Screening (HTS) method to directly quantify the reaction adducts. The reaction conditions have been optimized to minimize solubility issues, oxidative side reactions and increase the throughput of the assay while minimizing the reaction time, which are common issues with existing methods. Thirty-six chemicals previously classified with LLNA, DPRA or KeratinoSens™ were tested as a proof of concept. Preliminary results gave an estimated 82% accuracy, 78% sensitivity, 90% specificity, comparable to other in chemico methods such as Cys-DPRA. In addition to validated chemicals, six natural products were analyzed and a prediction of their sensitization potential is presented for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136581
1,5-Diazabicyclo[4.3.0]non-5-ene, 98%
C7H12N2
B59497
1-Bromobutane, ReagentPlus®, 99%
C4H9Br
237329
1-Chloro-2,4-dinitrobenzene, ≥99%
C6H3ClN2O4
180548
2,4-Heptadienal, technical grade, 90%
C7H10O
689203
3,4-Dihydroxy-3-cyclobutene-1,2-dione, Lonza quality, 98.50-101.00% (w/w) (HPLC)
C4H2O4
123447
3,4-Dihydroxy-3-cyclobutene-1,2-dione, 99%
C4H2O4
SML0415
Alantolactone, ≥98% (HPLC)
C15H20O2
P5648
β-Propiolactone, Grade II, ≥90%
C3H4O2
418099
Benzaldehyde, purified by redistillation, ≥99.5%
C7H6O
B1334
Benzaldehyde, ReagentPlus®, ≥99%
C7H6O
W212709
Benzaldehyde, ≥98%, FG, FCC
C7H6O
W212717
Benzaldehyde, natural, ≥98%, FCC, FG
C7H6O
B6630
Benzyl benzoate, ReagentPlus®, ≥99.0%
C14H12O2
B9550
Benzyl benzoate, meets USP testing specifications
C14H12O2
W213802
Benzyl benzoate, ≥99%, FCC, FG
C14H12O2
W213810
Benzyl benzoate, natural, ≥99%, FCC, FG
C14H12O2
W228613
Cinnamaldehyde, natural, ≥95%, FG
C9H8O
108197
Cinnamyl alcohol, 98%
C9H10O
W229407
Cinnamyl alcohol, ≥98%, FG
C9H10O
W229411
Cinnamyl alcohol, natural, 96%, FG
C9H10O
W358002
cis-6-Nonenal, ≥95%, FG
C9H16O
W230316
Citral, natural, ≥96%, FCC, FG
C10H16O
C83007
Citral, 95%
C10H16O
W230308
Citral, mixture of cis and trans, ≥96%, FG
C10H16O
SML0417
Costunolide, ≥97% (HPLC)
C15H20O2
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
02712
Cystamine dihydrochloride, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C4H12N2S2 · 2HCl
C8707
Cystamine dihydrochloride, BioXtra
C4H12N2S2 · 2HCl
C121509
Cystamine dihydrochloride, 96%
C4H12N2S2 · 2HCl