EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Protein-Protein Multilayer Oil-in-Water Emulsions for the Microencapsulation of Flaxseed Oil: Effect of Whey and Fish Gelatin Concentration.
PMID 26457588