EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Ring Opening Reactions through C-O Bond Cleavage Uniquely Adding Chemical Functionality to Boron Subphthalocyanine.
PMID 26457699
Abstract
We are reporting the unexpected reaction between bromo-boron subphthalocyanine (Br-BsubPc) and THF, 1,4-dioxane or γ-butyrolactone that results in the ring opening of the solvent and its addition into the BsubPc moiety. Under heating, the endocyclic C-O bond of the solvent is cleaved and the corresponding bromoalkoxy-BsubPc derivative is obtained. These novel alkoxy-BsubPc derivatives have remaining alkyl-bromides suitable for further functionalization. The alkoxy-BsubPcs maintain the characteristic strongly absorption in visible spectrum and their fluorescence quantum yields.