EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
New Signal Amplification Strategy Using Semicarbazide as Co-reaction Accelerator for Highly Sensitive Electrochemiluminescent Aptasensor Construction.
PMID 26457826
Abstract
A highly sensitive electrochemiluminescent (ECL) aptasensor was constructed using semicarbazide (Sem) as co-reaction accelerator to promote the ECL reaction rate of CdTe quantum dots (CdTe QDs) and the co-reactant of peroxydisulfate (S2O8(2-)) for boosting signal amplification. The co-reaction accelerator is a species that when it is introduced into the ECL system containing luminophore and co-reactant, it can interact with co-reactant rather than luminophore to promote the ECL reaction rate of luminophore and co-reactant; thus the ECL signal is significantly amplified in comparison with that in which only luminophore and co-reactant are present. In this work, the ECL signal probes were first fabricated by alternately assembling the Sem and Au nanoparticles (AuNPs) onto the surfaces of hollow Au nanocages (AuNCs) via Au-N bond to obtain the multilayered nanomaterials of (AuNPs-Sem)n-AuNCs for immobilizing amino-terminated detection aptamer of thrombin (TBA2). Notably, the Sem with two -NH2 terminal groups could not only serve as cross-linking reagent to assemble AuNPs and AuNCs but also act as co-reaction accelerator to enhance the ECL reaction rate of CdTe QDs and S2O8(2-) for signal amplification. With the sandwich-type format, TBA2 signal probes could be trapped on the CdTe QD-based sensing interface in the presence of thrombin (TB) to achieve a considerably enhanced ECL signal in S2O8(2-) solution. As a result, the Sem in the TBA2 signal probes could accelerate the reduction of S2O8(2-) to produce the more oxidant mediators of SO4(•-), which further boosted the production of excited states of CdTe QDs to emit light. With the employment of the novel co-reaction accelerator Sem, the proposed ECL biosensor exhibited ultrahigh sensitivity to quantify the concentration of TB from 1 × 10(-7) to 1 nM with a detection limit of 0.03 fM, which demonstrated that the co-reaction accelerator could provide a simple, efficient, and low-cost approach for signal amplification and hold great potential for other ECL biosensors construction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63768
3-Mercaptopropionic acid, ≥99.0% (HPLC)
C3H6O2S
M5801
3-Mercaptopropionic acid, ≥99%
C3H6O2S
439800
Cadmium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
CdCl2
202908
Cadmium chloride, 99.99% trace metals basis
CdCl2
655198
Cadmium chloride, technical grade
CdCl2
G5882
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
C5H8O2
G7651
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7776
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7526
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G6257
Glutaraldehyde solution, Grade II, 25% in H2O
C5H8O2
G6403
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
C5H8O2
340855
Glutaraldehyde solution, 50 wt. % in H2O
C5H8O2
49626
Glutaraldehyde solution, for electron microscopy, ~25% in H2O
C5H8O2
G4004
Glutaraldehyde solution, 25 wt. % in H2O
C5H8O2
W512303
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
C5H8O2
484431
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
C6H12O3
537543
Propylene glycol monomethyl ether acetate, 99%
C6H12O3
82300
Propylene glycol monomethyl ether acetate, ≥99.0% (GC)
C6H12O3