EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Upregulation of extracellular matrix metalloproteinase inducer promotes hypoxia-induced epithelial-mesenchymal transition in esophageal cancer.
PMID 26458866