EMAIL THIS PAGE TO A FRIEND
Biology Open
Generation of tissue-engineered small intestine using embryonic stem cell-derived human intestinal organoids.
PMID 26459240
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
269336
Peracetic acid solution, 32 wt. % in dilute acetic acid
C2H4O3