EMAIL THIS PAGE TO A FRIEND
Oncotarget
Aspirin, lysine, mifepristone and doxycycline combined can effectively and safely prevent and treat cancer metastasis: prevent seeds from gemmating on soil.
PMID 26459390
Abstract
Recent scientific advances have increased our understanding of the cancer metastatic complexities and provided further impetus for new combination therapies to prevent cancer metastasis. Here, we demonstrated that a combination (HAMPT) of aspirin, lysine, mifepristone and doxycycline can effectively and safely prevent cancer metastasis. The pharmaceutically-formulated HAMPT inhibited adhesion of cancer cells to either endothelial cells or extracellular matrix via down-regulating cell adhesion molecules ICAM-1 and α4-integrin. HAMPT inhibited the cloak effect by activated platelets on cancer cells, thereby interfering adhesion and invasion of cancer cells to the underlying stroma. At the effective concentration, HAMPT induced cancer cells into dormancy with minor inhibition on cell viability. Four-day pretreatment followed by 30-day oral administration of HAMPT (33.5-134 mg/kg) to the mice inoculated with cancer cells produced significant inhibition on cancer metastasis dose-dependently without marked side effects. Fifty-day rat toxicity study with HAMPT at doses (335-1340 mg/kg) 20-fold higher than its therapeutic dose produced no significant toxicity. Interestingly, the acute toxic death could not be reached at the maximum administrable dose (5 g/kg). This proof-of-concept study provides the first conceptual evidence that cancer metastasis can be controlled by using affordable old drugs to restrain circulating tumor cells from gemmating on the metastatic soil without the need for cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9891
Doxycycline hyclate, ≥98% (HPLC)
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O