EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
MicroRNA?328 directly targets p21?activated protein kinase 6 inhibiting prostate cancer proliferation and enhancing docetaxel sensitivity.
PMID 26459798