EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Development of an enantioselective assay for simultaneous separation of venlafaxine and O-desmethylvenlafaxine by micellar electrokinetic chromatography-tandem mass spectrometry: Application to the analysis of drug-drug interaction.
PMID 26460073