EMAIL THIS PAGE TO A FRIEND
International Journal of Clinical and Experimental Pathology
Caveolin-1 regulates cell apoptosis and invasion ability in paclitaxel-induced multidrug-resistant A549 lung cancer cells.
PMID 26464635