EMAIL THIS PAGE TO A FRIEND
International journal of clinical and experimental pathology
MicroRNA-204 inhibits cell proliferation in T-cell acute lymphoblastic leukemia by down-regulating SOX4.
PMID 26464665
Abstract
MicroRNAs (miRNAs) are a group of small non-coding RNAs that play important roles in the pathogenesis of human diseases by negatively regulating gene expression. The aim of this study was to explore the effect of miR-204 on cell proliferation migration and invasion in T-cell acute lymphoblastic leukaemia (T-ALL). miR-204 expression was determined in bone marrow samples from 32 leukemia patients and 32 healthy controls by quantitative real-time PCR (qRT-PCR). The effect of miR-204 on cell proliferation was evaluated by CCK8 assay, cell migration and invasion were evaluated by transwell migration and invasion assays, In addition, the regulation of SOX4 by miR-204 was evaluated by luciferase reporter assay and western blot. our results revealed that miR-204 was low expressed in T-ALL. Cell proliferation assay showed that the cell proliferation ability was inhibited by miR-204 mimics. Moreover, migration and invasion assay suggested that overexpression of miR-204 could significantly suppressed the migration and invasion ability of T-ALL cells. Luciferase reporter assay confirmed that miR-204 directly bound to the 3' untranslated region of SOX4, and western blot suggested that miR-204 inhibited the expression of SOX4 at the protein levels. Our findings indicated that miR-204 negatively regulates SOX4 and inhibited proliferation, migration and invasion of T-ALL cell lines. Thus, miR-204 might represent a potential therapeutic target for T-ALL intervention.