EMAIL THIS PAGE TO A FRIEND
Cell journal
Cell Attachment and Proliferation of Human Adipose-Derived Stem Cells on PLGA/Chitosan Electrospun Nano-Biocomposite.
PMID 26464814
Abstract
In this study, nano-biocomposite composed of poly (lactide-co-glycolide) (PLGA) and chitosan (CS) were electrospun through a single nozzle by dispersing the CS nano-powders in PLGA solution. The cellular behavior of human adipose derived stem cells (h-ADSCs) on random and aligned scaffolds was then evaluated. In this experimental study, the PLGA/CS scaffolds were prepared at the different ratios of 90/10, 80/20, and 70/30 (w/w) %. Morphology, cell adhesion and prolif- eration rate of h-ADSCs on the scaffolds were assessed using scanning electron microscope (SEM), 3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyl tetrazolium bromide (MTT) assay and trypan blue staining respectively. H-ADSCs seeded on the matrices indicated that the PLGA/CS composite matrix with aligned nanofibres and higher content of CS nano-powders gave significantly better performance than others in terms of cell adhesion and proliferation rate (P<0.05). We found that CS enhanced cell adhesion and proliferation rate, and aligned nanofibers guided cell growth along the longitudinal axis of the nanofibers, which would provide a beneficial approach for tissue engineering.