EMAIL THIS PAGE TO A FRIEND
Anesthesia and analgesia
The Efficacy of Eslicarbazepine Acetate in Models of Trigeminal, Neuropathic, and Visceral Pain: The Involvement of 5-HT1B/1D Serotonergic and CB1/CB2 Cannabinoid Receptors.
PMID 26465930
Abstract
Many clinical pain states that are difficult to treat share a common feature of sensitization of nociceptive pathways. Drugs that could normalize hyperexcitable neural activity (e.g., antiepileptic drugs) may be useful in relieving these pain states. Eslicarbazepine acetate (ESL) is a novel antiepileptic drug derived from carbamazepine/oxcarbazepine with a more favorable metabolic profile and potentially better tolerability. We examined the efficacy of ESL in models of inflammatory and neuropathic pain and the potential mechanism involved in its action. The antinociceptive effects of ESL were assessed in mice models of trigeminal (orofacial formalin test), neuropathic (streptozotocin-induced diabetic neuropathy model), and visceral pain (writhing test). The influence of 5-HT1B/1D serotonin receptor (GR 127935) and CB1 (AM251) and CB2 cannabinoid receptor (AM630) antagonists on the antinociceptive effect of ESL was tested in the model of trigeminal pain. ESL exhibited significant and dose-dependent antinociceptive effects in the second phase of the orofacial formalin test (P ≤ 0.011), in the tail-flick test in diabetic mice (P ≤ 0.013), and in the writhing test (P ≤ 0.003). GR 127935 (P ≤ 0.038) and AM251 and AM630 (P ≤ 0.013 for both antagonists) significantly inhibited the antinociceptive effect of ESL in a dose-related manner. ESL exhibited efficacy in models of trigeminal, neuropathic, and visceral pain. In the trigeminal pain model, the antinociceptive effect of ESL is, at least in part, mediated by 5-HT1B/1D serotonin and CB1/CB2 cannabinoid receptors. This study indicates that ESL could be useful in the clinical treatment of inflammatory and neuropathic pain.