EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part C, Methods
Functional Heart Valve Scaffolds Obtained by Complete Decellularization of Porcine Aortic Roots in a Novel Differential Pressure Gradient Perfusion System.
PMID 26467108