EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Field Efficacy of a Metarhizium anisopliae-Based Attractant-Contaminant Device to Control Ceratitis capitata (Diptera: Tephritidae).
PMID 26470297
Abstract
Biological control of Ceratitis capitata (Wiedemann) (Diptera: Tephritidae) using entomopathogenic fungi is being studied as a viable control strategy. The efficacy of a Metarhizium anisopliae (Metschnikoff) Sorokin (Hypocreales: Clavicipitaceae)-based attractant-contaminant device (ACD) to control C. capitata was evaluated in a medium-scale (40 ha) 2-yr field trial using a density of 24 ACD per ha. Results showed that this density was adequate to efficiently reduce fruitfly populations and that the inoculation dishes (IDs) needed replacing mid-season to provide protection for the entire season. In this study, fungal treatment was even more effective than conventional chemical treatment. Population dynamics in fungus-treated fields along with the infectivity study of field-aged IDs in the laboratory found that the ACD remained effective for at least 3 mo. The results suggest M. anisopliae-based ACD can be used to control C. capitata in the field. The implications of its use, especially as a tool in an integrated pest management program, are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCM-039
Malathion, vial of 1 g, analytical standard
C10H19O6PS2