EMAIL THIS PAGE TO A FRIEND
Food chemistry
Expression of pathogenesis-related (PR) genes in avocados fumigated with thyme oil vapours and control of anthracnose.
PMID 26471637
Abstract
Thyme oil (TO) fumigation (96μll(-1)) to cv. Hass and Ryan avocados significantly reduced anthracnose incidence compared to prochloraz and the untreated control. Also, enhanced activities of β-1,3-glucanase, chitinase were noted in both cultivars. TO fumigation induced the expression of both β-1,3-glucanase and chitinase genes in naturally infected fruit of both cultivars, during storage at 7 or 7.5°C for up to 21d and during subsequent simulated market shelf conditions at 20°C for 5d. However, the impact of TO fumigation on the β-1,3-glucanase gene expression was higher in both cultivars. Higher gene regulation and β-1,3-glucanase, chitinase activities were observed in cv. Ryan compared to Hass. Although TO fumigation significantly reduced anthracnose incidence in both naturally infected cultivars, the inhibitory effect was slightly higher in cv. Ryan than Hass. Thus, postharvest TO fumigation had positive effects on enhancing anthracnose disease resistance during storage and also gave a residual effect during the simulated shelf life.