EMAIL THIS PAGE TO A FRIEND
Basic research in cardiology
Activation of mitochondrial STAT-3 and reduced mitochondria damage during hypothermia treatment for post-cardiac arrest myocardial dysfunction.
PMID 26471891
Abstract
While therapeutic hypothermia improves the outcomes of individuals in cardiac arrest, the hemodynamic responses and mechanisms which underlie hypothermia-induced cardioprotection are not fully understood. Therefore, we investigated the mechanism by which induced hypothermia preserves cardiac function and protects against mitochondrial damage following cardiac arrest. Cardiac arrest was induced in adult male Wistar rats by asphyxiation for 8.5xa0min. Following resuscitation, the animals were randomly assigned to a hypothermia (32xa0°C) or normothermia (37xa0°C) group. Monitoring results showed that cardiac output at the fourth hour after resuscitation was significantly better in rats treated with hypothermia when compared to rats treated with normothermia (Pxa0