EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Bioprofiling of unknown antibiotics in herbal extracts: Development of a streamlined direct bioautography using Bacillus subtilis linked to mass spectrometry.
PMID 26472471
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46933
Citric acid, analytical standard
C6H8O7