EMAIL THIS PAGE TO A FRIEND
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer
Mechanisms of Acquired Resistance to AZD9291: A Mutation-Selective, Irreversible EGFR Inhibitor.
PMID 26473643
Abstract
AZD9291, a third-generation and mutation-selective epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor (TKI), is active against patients with EGFR-mutant non-small-cell lung cancer (NSCLC) who failed prior treatment with EGFR TKIs. However, acquired resistance to AZD9291 is inevitable. In this study, we identified the mechanisms of acquired resistance to AZD9291 in EGFR-mutant NSCLC. Four NSCLC patients with both an EGFR exon 19 deletion and the EGFR mutation after developing acquired resistance to first-generation EGFR TKIs received AZD9291 at doses of 20 to 80 mg/day in a phase I trial (NCT01802632). Paired tumor samples before and after treatment were obtained to evaluate EGFR modifications, alternative pathway activation, and histologic transformation. Genetic alterations were analyzed using Sanger sequencing, fluorescence in situ hybridization, real-time polymerase chain reaction, and targeted exome sequencing. All four patients achieved a partial response (median duration of response, 9 months [range, 9-11 months]) and subsequently showed resistance to AZD9291. EGFR-mutant clones depopulated AZD9291-resistant tumors to below 1% (baseline, 14%-36%) in three patients with progression: one with the loss of EGFR-double mutant clones and two accompanied by transformation to small-cell carcinoma and focal fibroblast growth factor receptor 1 (FGFR1) amplification, respectively. EGFR-mutant clones remained and the EGFR ligand was overexpressed in one patient with focal progression to AZD9291. Acquired resistance mechanisms of AZD9291 in patients with EGFR-mutant NSCLC who failed treatment with first-generation EGFR TKIs include the loss of EGFR-mutant clones plus alternative pathway activation or histologic transformation and EGFR ligand-dependent activation.