EMAIL THIS PAGE TO A FRIEND
Steroids
Determination of endocrine disruptors and endogenic androgens and estrogens in rat serum by high-performance liquid chromatography-tandem mass spectrometry.
PMID 26476180
Abstract
To simultaneously measure some targeted endocrine disruptors and several forms of sex hormones in rat serum, an accurate analytical procedure was developed. First, a comparison between a polymeric-based solid-phase extraction (SPE) and a micro-extraction by packed sorbent was performed to choose the optimal method to extract and concentrate the analytes: bisphenol A, atrazine, vinclozolin metabolite, testosterone, androstenedione, estrone, estradiol, estrone-sulfate and glucuronide and estradiol-sulfate and glucuronide. The analyses were then performed by high-performance liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) with electrospray ionisation in positive and negative modes. The protocol based on SPE was validated using the ICH/2005 guidelines. The validation demonstrated good performance in terms of linearity (R(2)>0.99), recovery (71-90%) and repeatability (relative standard deviation: 1-18%). The method was sensitive with LOQ comprised between 0.1 and 0.4 ng/ml for androgens and between 0.098 and 10.2 ng/ml for estrogens. The results obtained on the serum of rats exposed to the targeted endocrine disruptors showed the suitability of this analytical strategy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0251
Estrone 3-sulfate sodium salt, contains ~35% Tris as stabilizer
C18H21NaO5S