EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Novel tetracyclic benzo[b]carbazolones as highly potent and orally bioavailable ALK inhibitors: design, synthesis, and structure-activity relationship study.
PMID 26476749
Abstract
Four series of tetracyclic benzo[b]carbazolone compounds possessing more rotatable bonds and higher molecular flexibility were designed by either inserting a linker within the C8-side chain or by opening the middle ketone ring on the basis of compound 5 (Alectinib, CH5424802). Compound 15b was identified showing nearly identical high potency against both wild-type and the gatekeeper mutant ALK kinase (3.4 vs 3.9 nM). This compound has favorable PK profile with an oral bioavailability of 67.1% in rats. Moreover, compound 15b showed significant growth inhibition against ALK driven cancer cells and KARPAS-299 xenograft model.