EMAIL THIS PAGE TO A FRIEND
Nature communications
Enhancer repertoires are reshaped independently of early priming and heterochromatin dynamics during B cell differentiation.
PMID 26477271
Abstract
A widely accepted model posits that activation of enhancers during differentiation goes through a priming step prior to lineage commitment. To investigate the chronology of enhancer repertoire establishment during hematopoiesis, we monitored epigenome dynamics during three developmental stages representing hematopoietic stem cells, B-cell progenitors and mature B-cells. We find that only a minority of enhancers primed in stem cells or progenitors become active at later stages. Furthermore, most enhancers active in differentiated cells were not primed in earlier stages. Thus, the enhancer repertoire is reshaped dynamically during B-cell differentiation and enhancer priming in early stages does not appear to be an obligate step for enhancer activation. Furthermore, our data reveal that heterochromatin and Polycomb-mediated silencing have only a minor contribution in shaping enhancer repertoires during cell differentiation. Together, our data revisit the prevalent model about epigenetic reprogramming during hematopoiesis and give insights into the formation of gene regulatory networks.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48520-U
Chloroform, analytical standard
CHCl3
P9346
Liquified Phenol, ≥89.0%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
P1037
Phenol, for molecular biology
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
16018
Phenol, ≥96.0% (calc. on dry substance, T)
C6H6O
16016
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
C6H6O
W322318
Phenol, ≥99%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
185450
Phenol, unstabilized, ReagentPlus®, ≥99%
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
C6H6O
P4682
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3