EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Qualitative and quantitative analysis of N-acetyllactosamine and lacto-N-biose, the two major building blocks of human milk oligosaccharides in human milk samples by high-performance liquid chromatography-tandem mass spectrometry using a porous graphitic carbon column.
PMID 26477523