EMAIL THIS PAGE TO A FRIEND
Materials Science & Engineering. C, Materials For Biological Applications
Square wave voltammetric determination of paracetamol at chitosan modified carbon paste electrode: Application in natural water samples, commercial tablets and human urines.
PMID 26478288