EMAIL THIS PAGE TO A FRIEND
Pharmacological reports : PR
Antiarrhythmic activity of new 2-methoxyphenylpiperazine xanthone derivatives after ischemia/reperfusion in rats.
PMID 26481536
Abstract
We have previously shown significant prophylactic and therapeutic antiarrhythmic activity in adrenaline-induced arrhythmia, as well as α1-adrenolytic properties of new derivatives of xanthone. Herein, we investigated their antiarrhythmic activity in the model of ischemia/reperfusion in isolated hearts. Furthermore, we assessed antioxidant activity in biochemical studies. Antiarrhythmic activity in the model of ischemia/reperfusion in isolated perfused hearts was performed according to the Langendorff technique. Antioxidant activity was measured by lipid peroxidation level in tissue homogenate and in the FRAP assay. All studied compounds (MH-94, MH-99 and MH-105) showed significant antiarrhythmic activity in the model of ventricular arrhythmias associated with coronary artery occlusion and reperfusion. However, they did not demonstrate antioxidant effect, probably, because of the lack of free hydroxyl group(s) at a key position in the xanthone scaffold. The present study provides evidences for antiarrhythmic activity of some 2-methoxyphenylpiperazine derivatives of xanthone.