EMAIL THIS PAGE TO A FRIEND
Biomedical materials (Bristol, England)
Periodontal tissue regeneration with PRP incorporated gelatin hydrogel sponges.
PMID 26481592
Abstract
Gelatin hydrogels have been designed and prepared for the controlled release of the transforming growth factor (TGF-b1) and the platelet-derived growth factor (PDGF-BB). PRP (Platelet rich plasma) contains many growth factors including the PDGF and TGF-b1. The objective of this study was to evaluate the regeneration of periodontal tissue following the controlled release of growth factors in PRP. For the periodontal ligament cells and osteoblast, PRP of different concentrations was added. The assessment of DNA, mitochondrial activity and ALP activity were measured. To evaluate the TGF-β1 release from PRP incorporated gelatin sponge, amounts of TGF-β1 in each supernatant sample were determined by the ELISA. Transplantation experiments to prepare a bone defect in a rat alveolar bone were an implanted gelatin sponge incorporated with different concentration PRP. In DNA assay and MTT assay, after the addition of PRP to the periodontal ligament cells and osteoblast, the cell count and mitochondrial activity had increased the most in the group with the addition of 5  ×  PRP. In the ALP assay, after the addition of PRP to the periodontal ligament cells, the cell activity had increased the most in the group with the addition of 3  ×  PRP. In the transplantation, the size of the bone regenerated in the defect with 3  ×  PRP incorporated gelatin sponge was larger than that of the other group.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47829
D-(+) Glucose, analytical standard
C6H12O6
G7528
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
C6H12O6
G8270
D-(+)-Glucose, ≥99.5% (GC)
C6H12O6
G5767
D-(+)-Glucose, ACS reagent
C6H12O6
G7021
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
C6H12O6
G6152
D-(+)-Glucose, suitable for mouse embryo, ≥99.5% (GC)
C6H12O6
G5146
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
C6H12O6
49139
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
D9434
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
C6H12O6
G5882
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
C5H8O2
G7651
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7776
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7526
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G6257
Glutaraldehyde solution, Grade II, 25% in H2O
C5H8O2
G6403
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
C5H8O2
340855
Glutaraldehyde solution, 50 wt. % in H2O
C5H8O2
W512303
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
C5H8O2
G5417
Glycine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C2H5NO2
410225
Glycine, ACS reagent, ≥98.5%
C2H5NO2
G7126
Glycine, ReagentPlus®, ≥99% (HPLC)
C2H5NO2
50046
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
C2H5NO2
G8898
Glycine, for electrophoresis, ≥99%
C2H5NO2
G7403
Glycine, BioXtra, ≥99% (titration)
C2H5NO2
G8790
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
C2H5NO2
W328707
Glycine, 99%, FCC
C2H5NO2