EMAIL THIS PAGE TO A FRIEND
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment
Quantification of pyrrolizidine alkaloids in North American plants and honey by LC-MS: single laboratory validation.
PMID 26482059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17806
Senecionine, ≥95.0% (GC)
C18H25NO5