EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Identification of protein arginine N-methyltransferase 5 (PRMT5) as a novel interacting protein with the tumor suppressor protein RASSF1A.
PMID 26482848