EMAIL THIS PAGE TO A FRIEND
ACS Applied Materials & Interfaces
Trifunctional Polymeric Nanocomposites Incorporated with Fe3O4/Iodine-Containing Rare Earth Complex for Computed X-ray Tomography, Magnetic Resonance, and Optical Imaging.
PMID 26484385