EMAIL THIS PAGE TO A FRIEND
Nanoscale
Spontaneous and specific myogenic differentiation of human mesenchymal stem cells on polyethylene glycol-linked multi-walled carbon nanotube films for skeletal muscle engineering.
PMID 26486984
Abstract
This study explored the influence of polyethylene glycol-linked multi-walled carbon nanotube (PEG-CNT) films on skeletal myogenic differentiation of human mesenchymal stem cells (hMSCs). PEG-CNT films were prepared with nanoscale surface roughness, orderly arrangement of PEG-CNTs, high hydrophilicity and high mechanical strength. Notably, PEG-CNT films alone could direct the skeletal myogenic differentiation of hMSCs in the absence of myogenic induction factors. The quantitative real-time polymerase chain reaction (RT-PCR) showed that the non-induced hMSCs plated on the PEG-CNT films, compared to the negative control, presented significant up-regulation of general myogenic markers including early commitment markers of myoblast differentiation protein-1 (MyoD) and desmin, as well as a late phase marker of myosin heavy chain-2 (MHC). Corresponding protein analysis by immunoblot assays corroborated these results. Skeletal muscle-specific markers, fast skeletal troponin-C (TnC) and ryanodine receptor-1 (Ryr) were also significantly increased in the non-induced hMSCs on PEG-CNT films by RT-PCR. For these cells, the commitment to specific skeletal myoblasts was further proved by the absence of enhanced adipogenic, chondrogenic and osteogenic markers. This study elucidated that PEG-CNT films supported a dedicated differentiation of hMSCs into a skeletal myogenic lineage and can work as a promising material towards skeletal muscle injury repair.