EMAIL THIS PAGE TO A FRIEND
The Science of the Total Environment
Risk assessment of triazine herbicides in surface waters and bioaccumulation of Irgarol and M1 by submerged aquatic vegetation in Southeast Florida.
PMID 26490533