EMAIL THIS PAGE TO A FRIEND
Environmental science and pollution research international
A fusant of Amycolatopsis sp. M3-1 and Pseudomonas sp. Nai8 with high capacity of degrading novel pyrimidynyloxybenzoic herbicide ZJ0273 and naphthalene.
PMID 26490930
Abstract
ZJ0273 (propyl 4-(2-(4, 6-demethoxy pyrimidin-2-yloxy) benzylamino) benzoate) is a novel pyrimidynyloxybenzoic-based herbicide developed in China for oilseed crop. This study was aimed to construct new strains capable of degrading naphthalene and ZJ0273 by protoplast fusion between Amycolatopsis sp. M3-1 and Pseudomonas sp. Nai8. Eight recombinant strains were successfully produced, and the strains could simultaneously utilize ZJ0273 and naphthalene as the sole carbon and energy source, respectively. One of recombinant strains, MN6 with higher degrading efficiency, was chosen for further study. Under the condition of pH 7.0, 30 °C, ZJ0273 and naphthalene degradation percent by the recombinant strain MN6 could reach 65.10% (20 days) and 88.46% (48 h), respectively. According to the identified six metabolites (M1-M6) by LC-MS/MS, biodegradation pathway of ZJ0273 was proposed. ZJ0273 biodegradation catalyzed by the recombinant strain MN6 involved continuous biocatalytic reactions such as de-estering, hydrolysis, acylation, C-N cleavage, de-methyl, and ether cleavage reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105937
3,4,4′-Trichlorocarbanilide, 99%
C13H9Cl3N2O