EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences
Crystallization-induced dynamic resolution R-epimer from 25-OCH3-PPD epimeric mixture.
PMID 26491820