EMAIL THIS PAGE TO A FRIEND
Molecules (Basel)
Effect of Three Training Systems on Grapes in a Wet Region of China: Yield, Incidence of Disease and Anthocyanin Compositions of Vitis vinifera cv. Cabernet Sauvignon.
PMID 26492226