EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Stress Sensitivity Is Associated with Differential Accumulation of Reactive Oxygen and Nitrogen Species in Maize Genotypes with Contrasting Levels of Drought Tolerance.
PMID 26492235