EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
A Monolithic Hybrid Cellulose-2.5-Acetate/Polymer Bioreactor for Biocatalysis under Continuous Liquid-Liquid Conditions Using a Supported Ionic Liquid Phase.
PMID 26493884
Abstract
Mesoporous monolithic hybrid cellulose-2.5-acetate (CA)/polymer supports were prepared under solvent-induced phase separation conditions using cellulose-2.5-acetate microbeads 8-14 μm in diameter, 1,1,1-tris(hydroxymethyl)propane and 4,4'-methylenebis(phenylisocyanate) as monomers as well as THF and n-heptane as porogenic solvents. 4-(Dimethylamino)pyridine and dibutyltin dilaurate (DBTDL), respectively, were used as catalysts. Monolithic hybrid supports were used in transesterification reactions of vinyl butyrate with 1-butanol under continuous, supported ionic liquid-liquid conditions with Candida antarctica lipase B (CALB) and octylmethylimidazolium tetrafluoroborate ([OMIM(+) ][BF4 (-) ]) immobilized within the CA beads inside the polymeric monolithic framework and methyl tert-butyl ether (MTBE) as the continuous phase. The new hybrid bioreactors were successfully used in dimensions up to 2×30 cm (V=94 mL). Under continuous biphasic liquid-liquid conditions a constant conversion up to 96 % was achieved over a period of 18 days, resulting in a productivity of 58 μmol mg(-1) (CALB) min(-1) . This translates into an unprecedented turnover number (TON) of 3.9×10(7) within two weeks, which is much higher than the one obtained under standard biphasic conditions using [OMIM(+) ][BF4 (-) ]/MTBE (TON=2.7×10(6) ). The continuous liquid-liquid setup based on a hybrid reactor presented here is strongly believed to be applicable to many other enzyme-catalyzed reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
70956
1-Butyl-3-methylimidazolium hexafluorophosphate, ≥97.0% (HPLC)
C8H15F6N2P