EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Nontargeted Analysis Using Ultraperformance Liquid Chromatography-Quadrupole Time-of-Flight Mass Spectrometry Uncovers the Effects of Harvest Season on the Metabolites and Taste Quality of Tea (Camellia sinensis L.).
PMID 26494158