EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Notch increased vitronection adhesion protects myeloma cells from drug induced apoptosis.
PMID 26494298
Abstract
Notch signaling activation was found in many human cancers including multiple myeloma. It was previously reported that notch contributes to drug resistant of myeloma cells upon chemotherapy treatment, inhibition of notch by inhibitors helped to overcome drug resistance. However, the mechanism of notch developed drug resistance is remained to be fully illustrated. In the current study, we reported that Notch signaling activation up-regulated expression of integrin αvβ5 in myeloma cells companied with enhanced cells adhesion on vitronectin. Silencing Notch-1 receptor with siRNA or blocking cells with integrin αvβ5 antibody reduced myeloma cells adhesion on vitronectin, importantly, vitronectin mediated adhesion confers protection of myeloma cells from drug induced apoptosis. Thus, we revealed a novel mechanism of myeloma cells resistance to drug induced apoptosis. This study first connected Notch signaling, VTN adhesion and drug resistance together. Therefore, blocking αvβ5 receptor with antibody or knock down approach would be a novel promising strategy to treat MM.