EMAIL THIS PAGE TO A FRIEND
eLife
Attenuation of AMPK signaling by ROQUIN promotes T follicular helper cell formation.
PMID 26496200
Abstract
T follicular helper cells (Tfh) are critical for the longevity and quality of antibody-mediated protection against infection. Yet few signaling pathways have been identified to be unique solely to Tfh development. ROQUIN is a post-transcriptional repressor of T cells, acting through its ROQ domain to destabilize mRNA targets important for Th1, Th17, and Tfh biology. Here, we report that ROQUIN has a paradoxical function on Tfh differentiation mediated by its RING domain: mice with a T cell-specific deletion of the ROQUIN RING domain have unchanged Th1, Th2, Th17, and Tregs during a T-dependent response but show a profoundly defective antigen-specific Tfh compartment. ROQUIN RING signaling directly antagonized the catalytic α1 subunit of adenosine monophosphate-activated protein kinase (AMPK), a central stress-responsive regulator of cellular metabolism and mTOR signaling, which is known to facilitate T-dependent humoral immunity. We therefore unexpectedly uncover a ROQUIN-AMPK metabolic signaling nexus essential for selectively promoting Tfh responses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V8137
Anti-V5 antibody produced in rabbit, IgG fraction of antiserum