EMAIL THIS PAGE TO A FRIEND
PLoS Neglected Tropical Diseases
Polyanhydride Nanoparticle Delivery Platform Dramatically Enhances Killing of Filarial Worms.
PMID 26496201
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
655546
1,6-Bis(p-carboxyphenoxy)hexane, 90%
C20H22O6
D41007
1,6-Dibromohexane, 96%
C6H12Br2
M6762
1-Methyl-2-pyrrolidinone
C5H9NO
328634
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
C5H9NO
494496
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
C5H9NO
242799
1-Methyl-2-pyrrolidinone, spectrophotometric grade, ≥99%
C5H9NO
443778
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
C5H9NO
69118
1-Methyl-2-pyrrolidinone, puriss. p.a., ≥99.0% (GC)
C5H9NO
M79603
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
C5H9NO
282979
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
C7H12O2
A8380
5α-Androstan-17β-ol-3-one, ≥97.5%
C19H30O2
45726
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
C2H4O2
W200603
Acetic acid, natural, ≥99.5%, FG
C2H4O2
W200611
Acetic acid, ≥99.5%, FCC, FG
C2H4O2
ARK2183
Acetic acid, glacial, PharmaGrade, USP, JP, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C2H4O2
606766
Acetic acid-12C2, 99.9 atom % 12C
12C2H4O2
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
33214
Acetic anhydride, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99% (GC)
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
33085
Acetic anhydride
C4H6O3
L095004
Acetic anhydride
C4H6O3
439126
Acetone, for HPLC, ≥99.9%
C3H6O
00561
Acetone, for luminescence, ≥99.5% (GC)
C3H6O
40289
Acetone, semiconductor grade VLSI PURANAL (Honeywell 17617)
C3H6O
W332607
Acetone, ≥99%, meets FCC analytical specifications
C3H6O