EMAIL THIS PAGE TO A FRIEND
Biomaterials
3D brown adipogenesis to create Brown-Fat-in-Microstrands.
PMID 26496384
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9516
2-Propanol, BioReagent, for molecular biology, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
I7018
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
C10H14N4O2
I5879
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
C10H14N4O2
C5670
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
CaCl2
429759
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.99% trace metals basis
CaCl2
499609
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
CaCl2
C8106
Calcium chloride, meets USP testing specifications
CaCl2 · 2H2O
449709
Calcium chloride, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
CaCl2
21115
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
CaCl2
C7559
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
CHCl3
372978
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
CHCl3
288306
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
439142
Chloroform, for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
437581
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
480150
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
CHCl3
270636
Chloroform, for HPLC, ≥99.8%
CHCl3
48520-U
Chloroform, analytical standard
CHCl3
05-3433
Chloroform, for residue analysis, JIS 5000
CHCl3
05-3450
Chloroform, for HPLC
CHCl3
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D4902
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
C22H29FO5