EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Lack of Tissue Accumulation of Grape Seed Flavanols after Daily Long-Term Administration in Healthy and Cafeteria-Diet Obese Rats.
PMID 26496863
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1753
(−)-Epicatechin, ≥90% (HPLC)
C15H14O6
E4018
(−)-Epicatechin, ≥98% (HPLC), from green tea
C15H14O6
135011
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
C6H6O2
37580
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
C7H6O4
H20008
3-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
H49901
3-Hydroxyphenylacetic acid, ≥99%
C8H8O3
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
112003
Hippuric acid, 98%
C9H9NO3
P5561
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
C3H6O2
81910
Propionic acid, puriss. p.a., ≥99.5% (GC)
C3H6O2
W292400
Propionic acid, ≥99.5%, FCC, FG
C3H6O2
W292443
Propionic acid, ≥99.5%
C3H6O2
W292419
Propionic acid, natural, 99%, FG
C3H6O2
C9510
Pyrocatechol, ≥99%
C6H6O2
C3561
Pyrocatechol, plant cell culture tested, BioReagent, ≥99%, powder
C6H6O2
430749
Pyrocatechol, purified by sublimation, ≥99.5%
C6H6O2
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2
H36001
Vanillic acid, 97%
C8H8O4
W398802
Vanillic acid, ≥97%, FG
C8H8O4