EMAIL THIS PAGE TO A FRIEND
Oncotarget
A putative biomarker signature for clinically effective AKT inhibition: correlation of in vitro, in vivo and clinical data identifies the importance of modulation of the mTORC1 pathway.
PMID 26497682