EMAIL THIS PAGE TO A FRIEND
Oncotarget
MicroRNA expression profiling identifies miR-31-5p/3p as associated with time to progression in wild-type RAS metastatic colorectal cancer treated with cetuximab.
PMID 26497852
Abstract
The aim of our study was to investigate whether microRNAs (miRNAs) could serve as predictive biomarkers to anti-EGFR therapy (cetuximab, panitumumab) in patients with RAS wild-type (wt-RAS) metastatic colorectal cancer (mCRC). Historical cohort of 93 patients with mCRC (2006-2009) was included and further divided into exploratory and validation cohorts. MiRNAs expression profiling was performed on the exploratory cohort of 41 wt-KRAS mCRC patients treated with cetuximab to identify miRNAs associated with time to progression (TTP). The validation was performed on two independent cohorts: 28 patients of wt-RAS mCRC treated with cetuximab and 24 patients of wt-RAS mCRC treated with panitumumab. We identified 9 miRNAs with significantly different expression between responders and non-responders to cetuximab therapy (P ≤ 0.01). These 9 miRNAs were further evaluated in two independent cohorts of patients and miR-31-3p (P < 0.001) and miR-31-5p (P < 0.001) were successfully confirmed as strongly associated with TTP in wt-RAS mCRC patients treated with cetuximab but not panitumumab. When evaluated on the complete cohort of cetuximab patients (N = 69), miR-31-3p (HR, 5.10; 95% CI, 2.52-10.32; P < 0.001) and miR-31-5p (HR, 4.80; 95% CI, 2.50-9.24; P < 0.001) were correlated with TTP on the comparable level of significance. There was no difference in miR-31-5p/3p expression levels in RAS mutated and wild-type tumor samples. MiR-31-5p/3p are promising predictive biomarkers of cetuximab response in wt-RAS mCRC patients.